Learn More
Grainger Little Giant Cushion-Load Platform Truck, Steel, 1,200 lb, 1 ea
Catalog No. 502785156
Encompass_Preferred
This cushion-load platform truck is designed for efficient material handling, featuring a durable 12-gauge reinforced steel deck with a smooth, powder-coated finish. The low deck profile minimizes the lifting height required for loading and unloading bulky materials. It is equipped with four-ply pneumatic wheels that effectively absorb shock and reduce vibration, with adjustable inflation pressure to tailor the level of cushioning to specific load requirements.
  • Load capacity of 1,200 lb
  • Overall dimensions of 50 in L x 30 in W x 41-1/2 in H
  • Equipped with two rigid and two swivel 9 in pneumatic wheels
  • Includes a floor lock and a removable raised pipe handle
  • Solid, flush, all-welded steel construction
  • Assembled and ready for use
Catalog No. 50-278-5156 Supplier Grainger Supplier No. 19C192
This cushion-load platform truck is designed for efficient material handling, featuring a durable 12-gauge reinforced steel deck with a smooth, powder-coated finish. The low deck profile minimizes the lifting height required for loading and unloading bulky materials. It is equipped with four-ply pneumatic wheels that effectively absorb shock and reduce vibration, with adjustable inflation pressure to tailor the level of cushioning to specific load requirements.
  • Load capacity of 1,200 lb
  • Overall dimensions of 50 in L x 30 in W x 41-1/2 in H
  • Equipped with two rigid and two swivel 9 in pneumatic wheels
  • Includes a floor lock and a removable raised pipe handle
  • Solid, flush, all-welded steel construction
  • Assembled and ready for use