Learn More
Grainger Little Giant Platform Truck, Steel, 1,200 lb Capacity, 1 ea
Catalog No. 502784181
Encompass_Preferred
This cushion-load platform truck is designed for transporting heavy loads with reduced vibration and impact. Constructed with a 12 ga steel deck, it features a powder-coated finish for durability and a removable raised handle for versatile maneuverability. The unit is equipped with four 9 in. diameter pneumatic wheels to provide a cushioned ride over various floor surfaces.
  • 4-ply full pneumatic wheels absorb shock and reduce vibration
  • Adjustable tire inflation pressure allows for a customized level of cushion
  • Flush steel deck with smooth surface
  • Includes two rigid and two swivel casters for controlled handling
  • Sturdy construction with 1,200 lb load capacity
  • Removable raised handle provides ergonomic control
Catalog No. 50-278-4181 Supplier Grainger Supplier No. 19G872
This cushion-load platform truck is designed for transporting heavy loads with reduced vibration and impact. Constructed with a 12 ga steel deck, it features a powder-coated finish for durability and a removable raised handle for versatile maneuverability. The unit is equipped with four 9 in. diameter pneumatic wheels to provide a cushioned ride over various floor surfaces.
  • 4-ply full pneumatic wheels absorb shock and reduce vibration
  • Adjustable tire inflation pressure allows for a customized level of cushion
  • Flush steel deck with smooth surface
  • Includes two rigid and two swivel casters for controlled handling
  • Sturdy construction with 1,200 lb load capacity
  • Removable raised handle provides ergonomic control