Learn More
Grainger Little Giant Dual Handle Platform Truck, Steel, 3,600 lb, 12 ga, 1 ea
Catalog No. 502787208
Encompass_Preferred
This heavy-duty platform truck is designed for the efficient transport of large or heavy loads within industrial facilities. Built with a solid, 12 gauge steel deck and a durable powder-coated finish, it provides robust support for daily operations. The truck features handles at both ends for increased load retention and maneuverability, and it arrives fully assembled for immediate use.
  • Solid, 12 gauge steel deck with smooth surface
  • Load capacity of 3,600 lb.
  • Two removable pipe handles with two cross-braces for durability
  • Includes two rigid and two swivel polyurethane casters with 8 in. diameter wheels
  • Flush deck design for easy loading
  • Durable powder-coated finish on frame and deck
Catalog No. 50-278-7208 Supplier Grainger Supplier No. 19C193
This heavy-duty platform truck is designed for the efficient transport of large or heavy loads within industrial facilities. Built with a solid, 12 gauge steel deck and a durable powder-coated finish, it provides robust support for daily operations. The truck features handles at both ends for increased load retention and maneuverability, and it arrives fully assembled for immediate use.
  • Solid, 12 gauge steel deck with smooth surface
  • Load capacity of 3,600 lb.
  • Two removable pipe handles with two cross-braces for durability
  • Includes two rigid and two swivel polyurethane casters with 8 in. diameter wheels
  • Flush deck design for easy loading
  • Durable powder-coated finish on frame and deck