Learn More
Grainger PLATFORM TRUCK 72X30 RUBBER WH
Catalog No. 502784429
Encompass_Preferred
This general-purpose steel-deck platform truck is designed for heavy-duty material transport, featuring a sturdy all-welded construction. The unit provides a reliable solution for moving heavy loads in industrial environments.
  • Formed 12 gauge steel deck with extra reinforcement on the underside
  • Load capacity of 2,000 lb
  • Removable pipe handle featuring a 1-5/16 in. outer diameter and two cross braces
  • Four 6 in. diameter mold-on rubber caster wheels, including two swivel and two rigid configurations
  • Durable powder coated finish
  • Flush lip style deck surface
  • Assembled for immediate use
Catalog No. 50-278-4429 Supplier Grainger Supplier No. 19G875
This general-purpose steel-deck platform truck is designed for heavy-duty material transport, featuring a sturdy all-welded construction. The unit provides a reliable solution for moving heavy loads in industrial environments.
  • Formed 12 gauge steel deck with extra reinforcement on the underside
  • Load capacity of 2,000 lb
  • Removable pipe handle featuring a 1-5/16 in. outer diameter and two cross braces
  • Four 6 in. diameter mold-on rubber caster wheels, including two swivel and two rigid configurations
  • Durable powder coated finish
  • Flush lip style deck surface
  • Assembled for immediate use