Learn More
Grainger Little Giant Platform Truck, Steel, 2,400 lb Capacity, 1 ea
Catalog No. 502784456
Encompass_Preferred
This heavy-duty platform truck is an all-welded, single-end handle unit designed for transporting goods in demanding industrial environments. It features a robust 12 gauge steel deck with extra underside reinforcement and a durable powder-coated finish for long-lasting performance.
  • Solid 12 gauge steel deck with smooth, flush surface
  • Total load capacity of 2,400 lb
  • Four 8 in. mold-on rubber casters featuring a configuration of two rigid and two swivel wheels
  • Includes a floor lock and a removable raised pipe handle with two cross braces
  • Assembled unit construction
Catalog No. 50-278-4456 Supplier Grainger Supplier No. 19G870
This heavy-duty platform truck is an all-welded, single-end handle unit designed for transporting goods in demanding industrial environments. It features a robust 12 gauge steel deck with extra underside reinforcement and a durable powder-coated finish for long-lasting performance.
  • Solid 12 gauge steel deck with smooth, flush surface
  • Total load capacity of 2,400 lb
  • Four 8 in. mold-on rubber casters featuring a configuration of two rigid and two swivel wheels
  • Includes a floor lock and a removable raised pipe handle with two cross braces
  • Assembled unit construction