Learn More
Grainger Little Giant Platform Truck, Steel Deck, 2,400 lb Capacity, 1 ea
Catalog No. 502784185
Encompass_Preferred
This general-purpose platform truck features a robust all-welded steel construction designed for industrial and commercial material transport. The unit is built with a 12-gauge steel deck providing high durability and stability, supported by extra reinforcement on the underside to accommodate heavy loads.
  • Capacity of 2,400 lbs
  • Dimensions of 36 in L x 24 in W
  • Removable pipe handle with two cross braces for added durability
  • Four mold-on rubber casters in a 2 rigid and 2 swivel configuration
  • Caster wheels measure 8 in in diameter and 2 in in width
  • Powder-coated finish for corrosion resistance
  • Flush deck style
  • Assembled unit ready for immediate use
Catalog No. 50-278-4185 Supplier Grainger Supplier No. 19G864
This general-purpose platform truck features a robust all-welded steel construction designed for industrial and commercial material transport. The unit is built with a 12-gauge steel deck providing high durability and stability, supported by extra reinforcement on the underside to accommodate heavy loads.
  • Capacity of 2,400 lbs
  • Dimensions of 36 in L x 24 in W
  • Removable pipe handle with two cross braces for added durability
  • Four mold-on rubber casters in a 2 rigid and 2 swivel configuration
  • Caster wheels measure 8 in in diameter and 2 in in width
  • Powder-coated finish for corrosion resistance
  • Flush deck style
  • Assembled unit ready for immediate use