Learn More
Grainger Little Giant Narrow Steel Platform Truck, 2,000 Lb. Capacity, 60 in. x 16 in. Deck, 1 ea.
Catalog No. 502784427
Encompass_Preferred
This narrow steel-deck platform truck is designed for high-capacity material transport in confined aisles and busy workspaces. Built with a 12 ga steel deck, it provides a stable and reliable platform for heavy loads up to 2,000 lbs. Its six-wheel configuration allows for a zero-turn radius, providing maneuverability that is ideal for navigating tight environments.
  • Six-wheel design for enhanced maneuverability and zero-turn radius
  • High-capacity 2,000 lbs. load rating
  • Two rigid 8 in. center wheels and four 5 in. swivel corner-mounted casters
  • Two removable pipe handles extending 48 in. above the deck
  • Solid, smooth steel deck with powder-coated finish
  • Durable 12 ga steel construction
Catalog No. 50-278-4427 Supplier Grainger Supplier No. 19G786
This narrow steel-deck platform truck is designed for high-capacity material transport in confined aisles and busy workspaces. Built with a 12 ga steel deck, it provides a stable and reliable platform for heavy loads up to 2,000 lbs. Its six-wheel configuration allows for a zero-turn radius, providing maneuverability that is ideal for navigating tight environments.
  • Six-wheel design for enhanced maneuverability and zero-turn radius
  • High-capacity 2,000 lbs. load rating
  • Two rigid 8 in. center wheels and four 5 in. swivel corner-mounted casters
  • Two removable pipe handles extending 48 in. above the deck
  • Solid, smooth steel deck with powder-coated finish
  • Durable 12 ga steel construction