Learn More
Specifications
Specifications
| Chemical Name or Material | Platinizing Solution |
| CAS | 7732-18-5 |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| Grade | Laboratory |
| Packaging | Amber Glass Bottle |
| pH | ∼1 |
| Quantity | 120 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
