Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinizing Solution, For Electrical Conductivity and Resistivity, LabChem™

Catalog No. LC186807 Shop All LabChem Products
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC186807 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC186807 Supplier LabChem, Inc. Supplier No. LC186807
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Platinizing Solution
Color Orange-Yellow
Name Note For Electrical Conductivity and Resistivity
CAS Max % 96.7, 3, 0.3
CAS 18497-13-7, 6080-56-4
Linear Formula PtCl4·2HCl·6H2O
Density 1g/mL
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Soluble in water
Formula Weight 517.9
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1g/mL
Decomposition Information Hydrogen chloride; Lead oxide
Quantity 125 mL
Physical Form Liquid
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.