Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum, 0.3% on 2.7-3.3mm (0.11-0.13in) alumina pellets, reduced, Thermo Scientific Chemicals

Catalog No. AA3982614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3982614 25 g
AA3982622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3982614 Supplier Thermo Scientific Chemicals Supplier No. 03982614
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6-6-7440

Selective hydrogenation, gas purifications: e.g. Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Selective hydrogenation, gas purifications: e.g. Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-6-7440
MDL Number MFCD00011179

Specifications

Quantity 25 g
Chemical Name or Material Platinum, 0.3% on 2.7 to 3.3mm (0.11 to 0.13 in.) alumina pellets, Reduced
EINECSNumber 215-691-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.