Learn More
Platinum, 0.5% on granular carbon, reduced
Description
It is used as a catalytic agent in the processing of nitric acid, fertilizers, synthetic fibers, and a variety of other materials. The petroleum industry uses the metal in gauze form in crude oil refining.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Black |
| Appearance | Black granules |
| Quantity | 5 g |
| Odor | Odorless |
| Form | Granular |
| Solubility Information | Soluble in aqua regia. Insoluble in water,mineral and organic acids. |
| Chemical Name or Material | Platinum, 0.5% on granular carbon |
Safety and Handling
| Hazard Category | H228 |
| Hazard Statement | Keep container tightly closed. Take precautionary measures against static discharges. |
| Precautionary Statement | P210-P240-P241-P280-P370+P378q |
| DOTInformation | Hazard Class: 4.1; Packaging Group: III |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.