Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum, 1% on polyethyleneimine/SiO2 powder, Thermo Scientific™

Catalog No. p-7058876
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4340814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4340814 Supplier Thermo Scientific Chemicals Supplier No. 4340814
Only null left
Add to Cart

Reduction reactions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Soluble in aqua regia. Insoluble in water,mineral acids and organic acids.
Odor Odorless
Chemical Name or Material Platinum, 1% on polyethyleneimine/SiO2
Hazard Category H334
Hazard Statement GHS H Statement
H332-H335
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement P261-P285-P304+P341-P342+P311-P501c
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.