Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Platinum
Spécifications
Spécifications
| Melting Point | 1772°C |
| Boiling Point | 3827°C |
| Density | 21.45 g/cm3 (literature) |
| Quantity | 1 Ea. |
| Molecular Formula | Pt |
| Linear Formula | Pt |
| MDL Number | MFCD00011179 |
| Synonym | Platinum; PT000263 |
| Molecular Weight (g/mol) | 195.08 |
| Percent Purity | ≥99.99% |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'