Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Platinum

Numéro de catalogue. 267201400MG missing translation for 'brandsLinkText'
Encompass_Preferred
Modifier l'affichage
Click to view available options
Quantity:
400 mg
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
26-720-1400MG 400 mg
26-720-12G 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 26-720-1400MG Fournisseur Sigma Aldrich Code fournisseur 267201400MG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Ajouter au panier
Ajouter au panier

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Platinum

Spécifications

Melting Point 1772°C
Boiling Point 3827°C
Density 21.45 g/cm3 (literature)
Quantity 400 mg
Molecular Formula Pt
Linear Formula Pt
MDL Number MFCD00011179
Synonym Platinum element; Platinum gray
Molecular Weight (g/mol) 195.08
Percent Purity 99.99% trace metals basis
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.