Learn More
Strem, An Ascensus Company CAS# 7440-06-4. 25g. Platinum, 3% on activated wood carbon, reduced, 70% water wet paste (Escat™ 2931). MFCD00011179
SDP
Catalog No. 5090113273
Encompass
CAS# 7440-06-4. 25g. Platinum, 3% on activated wood carbon, reduced, 70% water wet paste (Escat™ 2931). MFCD00011179. Molecular Weight: . Molecular Formula: . Color/form: black pwdr. (d50=22 mum) Strem# 78-1614. http://www.strem.com/catalog/v/78-1614/
Catalog No. 50-901-13273 Supplier Strem, An Ascensus Company Supplier No. 78161425G
CAS# 7440-06-4. 25g. Platinum, 3% on activated wood carbon, reduced, 70% water wet paste (Escat™ 2931). MFCD00011179. Molecular Weight: . Molecular Formula: . Color/form: black pwdr. (d50=22 mum) Strem# 78-1614. http://www.strem.com/catalog/v/78-1614/