Learn More
Strem, An Ascensus Company CAS# 7440-06-4. 5g. Platinum, 5% on activated carbon, unreduced, 50% water wet paste (Escat™ 2441). MFCD00011179
SDP
Catalog No. 5090113270
Encompass
CAS# 7440-06-4. 5g. Platinum, 5% on activated carbon, unreduced, 50% water wet paste (Escat™ 2441). MFCD00011179. Molecular Weight: . Molecular Formula: . Color/form: black pwdr. (d50=18 mum) Strem# 78-1613. http://www.strem.com/catalog/v/78-1613/
Catalog No. 50-901-13270 Supplier Strem, An Ascensus Company Supplier No. 7816135G
CAS# 7440-06-4. 5g. Platinum, 5% on activated carbon, unreduced, 50% water wet paste (Escat™ 2441). MFCD00011179. Molecular Weight: . Molecular Formula: . Color/form: black pwdr. (d50=18 mum) Strem# 78-1613. http://www.strem.com/catalog/v/78-1613/