Learn More
ALADDIN SCIENTIFIC CORPORATION Platinum acetylacetonate, 1g
Catalog No. 5000082579
Encompass_Preferred
Used for photoactivated hydrosilylation reactions between vinyl and silane groups in the deposition of platinum metal and in the preparation of platinum nanotubes Precursor for FePt superlattice magnetic nanoparticles Pre-cursor for nano-materials Product classM-O Homogeneous Catalysts Acetylacetonate LigandsReaction typeHydrosilylationChemical propertiesChemical formulaC10H14O4PtEmpirical formula[Pt(acac)2]Molecular weight393 30MetalPtTheoretical metal content49Physical statepowderColoryellow
Catalog No. 50-000-82579 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. P1190251G
Used for photoactivated hydrosilylation reactions between vinyl and silane groups in the deposition of platinum metal and in the preparation of platinum nanotubes Precursor for FePt superlattice magnetic nanoparticles Pre-cursor for nano-materials Product classM-O Homogeneous Catalysts Acetylacetonate LigandsReaction typeHydrosilylationChemical propertiesChemical formulaC10H14O4PtEmpirical formula[Pt(acac)2]Molecular weight393 30MetalPtTheoretical metal content49Physical statepowderColoryellow