Learn More
Perkin Elmer US LLC Platinum Alloy Nebulizer Capillary Assembly with Platinum/Iridium Needle for AAnalyst
Catalog No. 502112771
Encompass_Preferred
This platinum alloy capillary assembly is etched with a P for identification. It is included in the Platinum Alloy Nebulizer Complete Assembly for AAnalyst (03030299). This capillary assembly is also compatible with Models 5100PC/5000/3300/3110/3100/1100(B)/2100/4100 AA series instruments.
Catalog No. 50-211-2771 Supplier Perkin Elmer US LLC Supplier No. 03030433
This platinum alloy capillary assembly is etched with a P for identification. It is included in the Platinum Alloy Nebulizer Complete Assembly for AAnalyst (03030299). This capillary assembly is also compatible with Models 5100PC/5000/3300/3110/3100/1100(B)/2100/4100 AA series instruments.