Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Platinum Black

Catalog No. 5207991G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-079-91G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-079-91G Supplier Sigma Aldrich Supplier No. 5207991G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Platinum black (low bulk density) can be electroplated on implantable microelectrode arrays (MEAs) to improve the impedence of the device. It can be used as a microelectrode for the formation of a microfluidic electrochemical sensor for glucose sensing applications. It can also be used as a photocathode for the production of hydrogen.

Specifications

CAS 121124-97-8
Melting Point 214°C to 217°C
Quantity 5 g
Molecular Formula C8H9BO4
Linear Formula HCOC6H3(OCH3)B(OH)2
MDL Number MFCD02093661
Synonym 3-Formyl-4-methoxybenzeneboronic acid; 5-Dihydroxyboryl-2-methoxybenzaldehyde
Molecular Weight (g/mol) 179.97 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.