Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum black, HiSPEC™ 1000

Catalog No. p-5547497
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1275503 1 g
AA1275506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1275503 Supplier Thermo Scientific Chemicals Supplier No. 01275503

CAS: 7440-06-4 | Pt | 195.08 g/mol

Platinum black is most commonly used for electrolyte fuel cell, microbial fuel cell, polymer electrolyte fuel cell and phosphoric acid fuel cells. It is used as a catalyst in hydrogenation reactions and a gas adsorbent. It can be coated on electrode surface by electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Appearance Dark gray powder
Quantity 1 g
Odor Odorless
Form Powder
Solubility Information Platinum black is insoluble in water and soluble in aqua regia.
Chemical Name or Material Platinum black
Hazard Category H228: Flammable solid.
EINECSNumber 231-116-1
RTECSNumber TP2160000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.