Learn More
Grainger Sensorex Platinum Contacting Conductivity Sensor, Epoxy, 6 In., PT1
Catalog No. 502768017
Encompass_Preferred
This platinum contacting conductivity sensor is designed for precise water quality analysis in industrial and light industrial applications. Constructed with an epoxy body and black-coated platinum measuring surface, it effectively monitors conductivity by applying a voltage potential across its electrodes. The sensor is optimized for use with the Con500 transmitter and provides reliable performance across a wide measurement range.
  • Designed for industrial and light industrial conductivity applications
  • Features an epoxy body with a black-coated platinum measuring surface
  • Supports a measurement range from 0 μS/cm to 10 μS/cm and 0 mS/cm to 199 mS/cm
  • Operating temperature range of 0°C to +70°C (32°F to 158°F)
  • Includes PT1 temperature sensor for accurate monitoring
  • Maximum pressure rating of 100 psig
  • Equipped with tinned leads for connection
Catalog No. 50-276-8017 Supplier Grainger Supplier No. 16Z058
This platinum contacting conductivity sensor is designed for precise water quality analysis in industrial and light industrial applications. Constructed with an epoxy body and black-coated platinum measuring surface, it effectively monitors conductivity by applying a voltage potential across its electrodes. The sensor is optimized for use with the Con500 transmitter and provides reliable performance across a wide measurement range.
  • Designed for industrial and light industrial conductivity applications
  • Features an epoxy body with a black-coated platinum measuring surface
  • Supports a measurement range from 0 μS/cm to 10 μS/cm and 0 mS/cm to 199 mS/cm
  • Operating temperature range of 0°C to +70°C (32°F to 158°F)
  • Includes PT1 temperature sensor for accurate monitoring
  • Maximum pressure rating of 100 psig
  • Equipped with tinned leads for connection