Learn More
Perkin Elmer US LLC Platinum Cover for Sample and Reference Cells, 1 ea.
Catalog No. 502104153
Encompass_Preferred
Each cover has two vent holes and is used for the Diamond DSC, Pyris 1 DSC, DSC 7, DSC 8000 and DSC 8500 sample holders. Platinum Cover for Sample and Reference Cells is for Pyris 1 DSC Autosampler and DSC 7 Robotic System. Cover location is different for the Pyris Diamond DSC. The entire length of the cover is 9.3 mm and a depth of 1.15 mm 0.05.
Catalog No. 50-210-4153 Supplier Perkin Elmer US LLC Supplier No. B0139122
Each cover has two vent holes and is used for the Diamond DSC, Pyris 1 DSC, DSC 7, DSC 8000 and DSC 8500 sample holders. Platinum Cover for Sample and Reference Cells is for Pyris 1 DSC Autosampler and DSC 7 Robotic System. Cover location is different for the Pyris Diamond DSC. The entire length of the cover is 9.3 mm and a depth of 1.15 mm 0.05.