Learn More
Sigma Aldrich Fine Chemicals Biosciences Platinum crucible lid fits1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5621061EA
Encompass
Platinum labware has excellent resistance to acids such as acetic hydrochloric hydrofluoric hydroiodic nitric selenic and sulfuric and to oxidation at high temperatures. Crucible lids of platinum can be cleaned by boiling in a suitable solvent.For the removal of substances which have alloyed with the surface scour with alumina powder.
Catalog No. 50-290-8876