Learn More
Cole-Parmer Platinum Crucible, wide form, 50 mL, 1/ea
Supplier:  Cole-Parmer 1783082
Encompass
These crucibles offer high temperature strength and high melting point. Platinum offers oxidation resistance in air and oxygen; and corrosion resistance in acids and melted salts. Protect your investment with platinum tongs that reduce possible damage and scratches to platinum crucible when hot. Manufactured in standard form. Lids, crucibles, and tongs sold separately.
Catalog No. NC1588971