Learn More
Sigma Aldrich Fine Chemicals Biosciences Platinum crucible with rei1EA
Catalog No. 502908716
Encompass
Platinum labware has excellent resistance to acids such as acetic hydrochloric hydrofluoric hydroiodic nitric selenic and sulfuric and to oxidation at high temperatures. These platinum crucibles can be cleaned by boiling in a suitable solvent for the removal of substances which have alloyed with the surface scour with alumina powder. Having a reinforced rim means that the rim of the crucible is thicker than the walls to make it stronger this is where the crucible can be gripped if tongs are used during the handling process.
Catalog No. 50-290-8716 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6853051EA
Platinum labware has excellent resistance to acids such as acetic hydrochloric hydrofluoric hydroiodic nitric selenic and sulfuric and to oxidation at high temperatures. These platinum crucibles can be cleaned by boiling in a suitable solvent for the removal of substances which have alloyed with the surface scour with alumina powder. Having a reinforced rim means that the rim of the crucible is thicker than the walls to make it stronger this is where the crucible can be gripped if tongs are used during the handling process.