Learn More
Electron Microscopy Sciences Platinum Crucible Lid, 1 ml
SDP
Catalog No. 5032933
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 1 ml
Catalog No. 50-329-33 Supplier Electron Microscopy Sciences Supplier No. 665011
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 1 ml