Learn More
Electron Microscopy Sciences Platinum Crucible, 1 ml, 13 mm height, 12.0 mm O.D., 7.2 mm base
SDP
Supplier:  Electron Microscopy Sciences 665001
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 1 ml
Catalog No. 50-329-23
 Restricted Item