Learn More
Electron Microscopy Sciences Platinum Crucible, 10 ml, 29 mm height, 26.0 mm O.D., 15.5 mm base
SDP
Catalog No. 5032921
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 10 ml
Catalog No. 50-329-21 Supplier Electron Microscopy Sciences Supplier No. 665042
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 10 ml