Learn More
Electron Microscopy Sciences Platinum Crucible Lid, 10 ml
SDP
Catalog No. 5032910
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 10 ml
Catalog No. 50-329-10 Supplier Electron Microscopy Sciences Supplier No. 665052
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 10 ml