Learn More
Electron Microscopy Sciences Platinum Crucible, 15 ml, 33 mm height, 29.0 mm O.D., 17.5 mm base
SDP
Catalog No. 5032900
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 15 ml
Catalog No. 50-329-00 Supplier Electron Microscopy Sciences Supplier No. 665044
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 15 ml