Learn More
Electron Microscopy Sciences Platinum Crucible, 15 ml, 27 mm height, 32.0 mm O.D., 19.5 mm base
SDP
Catalog No. 5032901
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 15 ml
Catalog No. 50-329-01 Supplier Electron Microscopy Sciences Supplier No. 665045
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 15 ml