Learn More
Electron Microscopy Sciences Platinum Crucible Lid, 15 ml
SDP
Catalog No. 5032914
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 15 ml
Catalog No. 50-329-14 Supplier Electron Microscopy Sciences Supplier No. 665055
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 15 ml