Learn More
Electron Microscopy Sciences Platinum Crucible, 20 ml, 31 mm height, 37.0 mm O.D., 22.5 mm base
SDP
Catalog No. 5032903
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 20 ml
Catalog No. 50-329-03 Supplier Electron Microscopy Sciences Supplier No. 665047
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 20 ml