Learn More
Electron Microscopy Sciences Platinum Crucible, 20 ml, 36 mm height, 32.5 mm O.D., 18.0 mm base
SDP
Supplier:  Electron Microscopy Sciences 665046
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 20 ml
Catalog No. 50-329-02