Learn More
Electron Microscopy Sciences Platinum Crucible Lid, 20 ml
SDP
Supplier:  Electron Microscopy Sciences 665057
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 20 ml
Catalog No. 50-329-16