Learn More
Electron Microscopy Sciences Platinum Crucible Lid, 20 ml
SDP
Catalog No. 5032915
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 20 ml
Catalog No. 50-329-15 Supplier Electron Microscopy Sciences Supplier No. 665056
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 20 ml