Learn More
Electron Microscopy Sciences Platinum Crucible, 25 ml, 34 mm height, 41 mm O.D., 25 mm base
SDP
Catalog No. 5032904
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 25 ml
Catalog No. 50-329-04 Supplier Electron Microscopy Sciences Supplier No. 665048
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 25 ml