Learn More
Electron Microscopy Sciences Platinum Crucible, 25 ml, 39 mm height, 35.0 mm O.D., 21.0 mm base
SDP
Catalog No. 5032905
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 25 ml
Catalog No. 50-329-05 Supplier Electron Microscopy Sciences Supplier No. 665049
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 25 ml