Learn More
Electron Microscopy Sciences Platinum Crucible Lid, 25 ml
SDP
Catalog No. 5032918
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 25 ml
Catalog No. 50-329-18 Supplier Electron Microscopy Sciences Supplier No. 665059
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 25 ml