Learn More
Electron Microscopy Sciences Platinum Crucible Lid, 3 ml
SDP
Catalog No. 5032926
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 3 ml
Catalog No. 50-329-26 Supplier Electron Microscopy Sciences Supplier No. 665013
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 3 ml