Learn More
Electron Microscopy Sciences Platinum Crucible, 3 ml, 19 mm height, 17.0 mm O.D., 10.0 mm base
SDP
Supplier:  Electron Microscopy Sciences 665003
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 3 ml
Catalog No. 50-329-24
 Restricted Item