Learn More
Electron Microscopy Sciences Platinum Crucible Lid, 30 ml, 41 mm height, 37.5 mm O.D., 22.5 mm base
SDP
Catalog No. 5032906
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 30 ml
Catalog No. 50-329-06 Supplier Electron Microscopy Sciences Supplier No. 6650510
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 30 ml