Learn More
Electron Microscopy Sciences Platinum Crucible, 35 ml, 36 mm height, 43.5 mm O.D., 26.5 mm base
SDP
Catalog No. 5032928
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 35 ml
Catalog No. 50-329-28 Supplier Electron Microscopy Sciences Supplier No. 6650411
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 35 ml