Learn More
Electron Microscopy Sciences Platinum Crucible Lid, 35 ml, 36 mm height, 43.5 mm O.D., 26.5 mm base
SDP
Catalog No. 5032907
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 35 ml
Catalog No. 50-329-07 Supplier Electron Microscopy Sciences Supplier No. 6650511
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 35 ml