Learn More
Electron Microscopy Sciences Platinum Crucible Lid, 40 ml, 46 mm height, 41.5 mm O.D., 25.0 mm base
SDP
Supplier:  Electron Microscopy Sciences 6650513
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 40 ml
Catalog No. 50-329-08
 Restricted Item