Learn More
Electron Microscopy Sciences Platinum Crucible, 5 ml, 22 mm height, 20.5 mm O.D., 12.0 mm base
SDP
Catalog No. 5032925
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 5 ml
Catalog No. 50-329-25 Supplier Electron Microscopy Sciences Supplier No. 665005
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 5 ml