Learn More
Electron Microscopy Sciences Platinum Crucible Lid, 5 ml
SDP
Catalog No. 5032920
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 5 ml
Catalog No. 50-329-20 Supplier Electron Microscopy Sciences Supplier No. 665015
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 5 ml