Learn More
Electron Microscopy Sciences Platinum Crucible Lid, 50 ml, 49 mm height, 44.0 mm O.D., 26.5 mm base
SDP
Catalog No. 5032919
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 50 ml
Catalog No. 50-329-19 Supplier Electron Microscopy Sciences Supplier No. 6650515
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 50 ml