Learn More
Electron Microscopy Sciences Platinum Crucible Lid, 50 ml, 41 mm height, 49.0 mm O.D., 30.0 mm base
SDP
Catalog No. 501899203
Encompass_Preferred
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 50 ml
Catalog No. 50-189-9203 Supplier Electron Microscopy Sciences Supplier No. 6650514
Made from platinum, resulting in excellent resistance to chemical attacks by acids and fusion mixtures.
Capacity: 50 ml