Learn More
Accuflow Systems Platinum-cured Silicone Tubing
Catalog No. 502995684
Encompass_Preferred
Purity One™ OneSil 65D biopharmaceutical grade silicone tubing has been formulated for demanding and critical fluid transfer applications in biopharmaceutical processing. This general use silicone is moldable for single-use assemblies, has higher pressure ratings and better kink resistance than traditional 50D silicone tubing, and has a large temperature range, making it an excellent choice for applications such as sampling, filtration, sterile filling and transfer, and buffer and media prep. Full validation guide and USP <665> Extractables testing available.
Catalog No. 50-299-5684 Supplier Accuflow Systems Supplier No. ONESIL6500630188
Purity One™ OneSil 65D biopharmaceutical grade silicone tubing has been formulated for demanding and critical fluid transfer applications in biopharmaceutical processing. This general use silicone is moldable for single-use assemblies, has higher pressure ratings and better kink resistance than traditional 50D silicone tubing, and has a large temperature range, making it an excellent choice for applications such as sampling, filtration, sterile filling and transfer, and buffer and media prep. Full validation guide and USP <665> Extractables testing available.